Mini-Dolly Folding Cart
Mini-Dolly Folding Cart
Actual product may vary
Mini-Dolly Folding Cart
Mini-Dolly Folding Cart
Mini-Dolly Folding Cart
Folds to a compact size, Fisherbrand Mini-Dolly Folding Cart is made from high-impact ABS plastic with chrome-plated steel durability.
Have Questions?
Catalog NumberMaterial
CTL04-681ABS Plastic, Chrome-plated Steel
Catalog number CTL04-681
Price (SGD)
499.00
EA
Add to cart
Material:
ABS Plastic, Chrome-plated Steel
Price (SGD)
499.00
EA
Add to cart
Folds to a compact size, Fisherbrand Mini-Dolly Folding Cart is made from high-impact ABS plastic with chrome-plated steel durability.
  • Load capacity: 265 lb. (120kg)
  • Constructed of ABS plastic and chrome-plated steel with low, nonskid platform
  • Eliminates lifting required by high-shelf carts
  • Easy-roll wheels swivel for added maneuverability
  • Plastic guard on four sides protects walls
  • Easy-grip handle (34.5 in. high) ensures a firm hold
  • Folds for storage
Specifications
TypeMini-dolly Folding Cart
Load Bearing Capacity (English)265 lb.
Load Bearing Capacity (Metric)120 kg
MaterialABS Plastic, Chrome-plated Steel
For Use With (Application)For platform cart to move chemicals, carboys, computer paper, filing boxes and scientific equipment, storage purpose
Height (Metric)34.5 in. high handle
IncludesThree bungee cords, easy roll wheels swivel, Plastic guard on four sides
Length (English) Shelf25.5 in.
Length (Metric) Shelf64.7 cm
Weight (English)20 lb.
Weight (Metric)9 kg
Width (English)17.75 in.
Width (Metric)45 cm
Unit SizeEA