Traceable™ Stopwatch/Repeat Timer
Traceable™ Stopwatch/Repeat Timer
Actual product may vary
Traceable™ Stopwatch/Repeat Timer
Ideal for an analysis that must be noted at routine intervals
Have Questions?
Catalog NumberIncludes
CTL02-261-961Lanyard
Catalog number CTL02-261-961
Price (SGD)
-
Includes:
Lanyard
Ideal for an analysis that must be noted at routine intervals. Fisher Scientific™ Traceable™ Stopwatch/Repeat Timer provides stopwatch functions of single-action, time-in/-out, cumulative-split, interval-split, eight memories, and continuous timing.
Set countdown time from 10 hours to 1/10 of a second. When zero is reached, timer alarms and repeats countdown from the programmed time. Repeat cycle automatically continues until stopped. Display count box shows how many times the cycle has been repeated. LCD screen continually displays time remaining to zero.
  • Universal stopwatch provides repeat function of countdown alarm timing
  • ABS plastic shockproof case
  • Timing range is 10 hours to 1/10 of a second
  • Resolution: 0.01 second
  • Accuracy: 0.001%
  • Size: 6 x 6 x 1.3 cm (2.375 x 2.375 x 0.5 in.)
  • Weight: 1.75 oz.
Specifications
Product TypeDigital Stopwatch/Timer
Time Range10 hr.
AlarmsAudible
Accuracy1e-05
Resolution0.01 sec.
DisplayLCD
Weight (English)1.75 oz.
Weight (Metric)0.049 kg
IncludesLanyard
For Use With (Application)Analysis that must be noted at routine intervals
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEach