Battery Charger
Actual product may vary
CleanSpace
Battery Charger
The CleanSpace Battery Charger is specifically designed to re-charge the internal lithium polymer battery of the CleanSpace Power Units. The battery charger has two leads and interchangeable plugs for use in different countries.
Have Questions?
Catalog number CSEPAF-1101
Price (SGD)
84.00
Each
Price (SGD)
84.00
Each
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Multinational interchangeable plugs
  • Dual leads for charging two PAPR power units simultaneously
Specifications
Product TypeCharger
For Use With (Equipment)PAF-0034, PAF-0060, PAF-0070
Unit SizeEach