Full Face PAPR Mask
Full Face PAPR Mask
Actual product may vary
CleanSpace
Full Face PAPR Mask
Full face mask is suitable for heavy, long term work, or exposure. Designed for use with the CleanSpace™ Ultra and EX PAPR systems.
Have Questions?
Catalog NumberSize
CSEPAF-1014Medium/Large
Catalog number CSEPAF-1014
Price (SGD)
526.00
EA
Add to cart
Size:
Medium/Large
Price (SGD)
526.00
EA
Add to cart
  • Used with the revolutionary CleanSpace™PAPR: lightweight, no hoses or belts
  • Wide panoramic visor
  • Hard scratch and solvent resistant coating
  • Suitable for long periods of wear
  • Positive pressure airflow and inner Orinasal mask prevents fogging
  • Easy breathing and no resistance even during heavy exertion
  • Visor seal silicone design seals to the face for optimum protection and comfort.
  • Inner Orinasal mask with unique clip-in clip-out design is easily removed for cleaning
  • Mylar speech diaphragm for clear communication on the job
  • Age resistant materials
  • PortaCount testing enabled for quantitative mask fit testing (Sold separately)
  • Spectacle kit: Fully adjustable for all face sizes, wide frames to fit large lenses for peripheral vision; engineered stability during vigorous activity (Sold separately)
  • Tear-Offs: Visor protection against scratches (Sold separately)
  • Hard moulded polycarbonate visor; Silicone face seal and Inner Orinasal Mask cushion. Nylon mask clips; Mylar speech diaphragm. EPDM rubber harness; impact modified reinforced polypropylene exhalation valve housing; silicone O-rings
Specifications
Certifications/ComplianceAS/NZS1716: 2012
Certifications/Compliance (Europe)EN 12942
Product TypePAPR Full Face Mask
SizeMedium/Large
Facepiece/Headtop ModelPAPR full face mask only
For Use With (Equipment)PAF-1070
ModelPAF-1014
Storage RequirementsStore at -10°C to 55°C
Unit SizeEA