PAPR Half Mask
Actual product may vary
CleanSpace
PAPR Half Mask
Half masks clip quickly onto PAPR power units and easily unclip for cleaning. Specifically designed to be used with the CleanSpace™ 2, Ultra and EX PAPR systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
CSEPAF-1010Medium
CSEPAF-0033Small
CSEPAF-0027Large
Catalog number CSEPAF-1010
Price (SGD)
229.00
Each
Size:
Medium
Price (SGD)
229.00
Each
  • Compatible with ALL CleanSpace™PAPR power units
  • Used with the revolutionary CleanSpace™PAPR: lightweight, no hoses or belts
  • Available in 3 sizes to facilitate a secure seal and a comfortable fit
  • Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
  • Easy removal for cleaning
  • Transparent and facilitates communication
  • Clip quickly onto the power units and are easily unclipped for cleaning
  • Soft silicone that readily moulds to the face and does not cause allergic dermatitis
Specifications
Certifications/ComplianceAS/NZS1716: 2012
Certifications/Compliance (Europe)EN 12942
Certifications (EN)EN 12942
Product TypePAPR Half Mask
SizeMedium
Facepiece/Headtop ModelPAPR half mask only
For Use With (Application)Mining, welding, manufacturing, smelting, construction, recycling plants, emergency services, agriculture, processing plants, grinding
For Use With (Equipment)PAF-1034, PAF-2034
IP RatingIP66/67
ModelPAF-1010
Storage RequirementsStore at -10°C to 55°C
Dimensions (L x W x H)170 x 130 x 70 mm
Weight (Metric)125 g
Unit SizeEach