Battery-powered, positive pressure air-purifying respirator with an IP Rating 66. Suitable for decontamination procedures and applications where there is an abundance of dust and water. Can be used for particulate and gas applications.
Have Questions?
Catalog number CSEPAF-0070
Price (SGD)
1,368.00
Special offer
Ends: 31-Mar-2026
2,137.00
Save 769.00 (36%)
Each
Add to cart
Price (SGD)
1,368.00
Special offer
Ends: 31-Mar-2026
2,137.00
Save 769.00 (36%)
Each
Add to cart
IP Rating 66 – resistant to dust and liquid ingress
Suitable for decontamination showers
Light weight and comfortable
AirSensit™ System for mask pressure control and breath responsive airflow
One-button system and standby and auto-start/stop mode for quick don/doffing
Easy to use support harness
Altitude compensated filter blocked detection
Long operating time: up to 8 hours and quick battery recharging: < 2 hours