Suitable for protection against airborne particulates (dust, mists and fumes). Designed to remove course particles and enhance the life of the Particulate P3 Filter (PAF-0035. The Protective Cover and Pre-Filter should be changed after each use.
Have Questions?
Catalog number CSEPAF-0058
Price (SGD)
73.00
Each
Price (SGD)
73.00
Each
Used with the revolutionary CleanSpace - a light weight PAPR with no hoses/belts
Materials: Spun polymer fibers
Easy and quickly fitted and removed from the power unit