The CleanSpace Combined Filter TM3 ABEK1 P SL R is suitable for protection against airborne particulate (dust, mists and fumes), organic, inorganic gases or vapours, solvents, ammonia and sulphur dioxide (boiling point >65C).
Have Questions?
Catalog number CSEPAF-0052
Price (SGD)
105.00
Special offer
Ends: 31-Mar-2026
114.00
Save 9.00 (8%)
Each
Add to cart
Price (SGD)
105.00
Special offer
Ends: 31-Mar-2026
114.00
Save 9.00 (8%)
Each
Add to cart
Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
Suitable for protection against airborne particulate (dust, mists and fumes organic, inorganic gases or vapors, ammonia and sulphur dioxide (boiling point > 65 °C)
Materials: Fiberglass particulate media, activated carbon and plastic casing, silicone seal