Filter Adaptor
Actual product may vary
CleanSpace Respirators
Filter Adaptor
Filter adaptor must be in placed when using the CleanSpace™ large case filter.
Have Questions?
Catalog NumberFor Use With (Equipment)
CSEPAF-0038PAF-1034, PAF-1070
Catalog number CSEPAF-0038
Price (SGD)
93.00
Each
For Use With (Equipment):
PAF-1034, PAF-1070
Price (SGD)
93.00
Each
  • Compatible with the CleanSpace™PAPR power units PAF-1034 and PAF-2034
  • Suitable for all CleanSpace™large filters: PAF-1003
  • Materials: Plastic casing, silicone seal and steel fasteners and stainless steel springs.
  • Easy and quickly fitted and removed from the power unit
Specifications
Certifications/ComplianceAS/NZS1716: 2012, EN 12942
DisposableReusable
Product TypeFilter Adapter
For Use With (Equipment)PAF-1034, PAF-1070
ModelPAF-0038
Storage RequirementsStore at -10°C to +55°C
Dimensions (L x W x H)180 x 110 x 80 mm
Weight (Metric)113 g
Unit SizeEach