Particulate Filter P3 TM3 P SL R
Actual product may vary
CleanSpace
Particulate Filter P3 TM3 P SL R
The CleanSpace Particulate Filter P3 TM3 P SL R is suitable for protection against airborne particulates (dust, mists and fumes).
Have Questions?
Catalog number CSEPAF-0035
Price (SGD)
134.00
Pack
Price (SGD)
134.00
Pack
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Suitable for protection against airborne particulate (dust, mists and fumes).
  • Materials: Fiberglass particulate media and plastic casing, silicone seal
  • Easy and quickly fitted and removed from the power unit
  • Pack of 3 (PAF-0035) or Pack of 24 (PAF-0059)
Specifications
Product TypeFilter
Unit SizePack