Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Fit Testing Kits and Supplies
▸
Half Mask Adaptor
Actual product may vary
CleanSpace
Half Mask Adaptor
Half mask adaptor is equipped for the front of the mask to enable quantitative fit testing.
Have Questions?
Catalog Number
For Use With (Equipment)
CSEPAF-0025
PAF-1010, PAF-0027, PAF-0033
Catalog number
CSEPAF-0025
Price (SGD)
266.00
Each
Add to cart
For Use With (Equipment):
PAF-1010, PAF-0027, PAF-0033
Price (SGD)
266.00
Each
Add to cart
Used with the revolutionary CleanSpace™ Respirators: light weight, no hoses or belts
Used with the CleanSpace™ Half Masks only
Quick and easy to install
Fits with the TSI PortaCount equipment
Easy to wash and quick drying
Specifications
Product Type
Half Mask Adapter
For Use With (Application)
Quantitative Fit Testing
For Use With (Equipment)
PAF-1010, PAF-0027, PAF-0033
Model
PAF-0025