Steri-Plus Exhalation Valve Filter for Source Control
Actual product may vary
CleanSpace
Steri-Plus Exhalation Valve Filter for Source Control
Disposable exhalation valve filter is utilised for source control in sterile settings where filtering of the wearers exhaled air is required.
Have Questions?
Catalog number CSECS3039
Price (SGD)
66.00
Pack
Price (SGD)
66.00
Pack
The world's first PAPR exhalation valve filter for source control. Developed to filter the air exhaled by the wearer and consequently protect those not wearing a respiratory protection system.
  • Filtration efficiency of 97% for particles 0.3 μm and above
  • Approved for use with CleanSpace™ respirators and half masks
  • Reusable case is compatible with standard disinfection and sterilization protocols
  • Easy to fit and clean
  • Cost-effective
  • NIOSH and CE Mark approved
  • Suitable for use in patient care settings and laboratory or pharmaceutical clean rooms
  • Provides reliable source control for CleanSpace™ respirators when used with the half mask
  • Exhalation valve filter media is nonwoven electrostatic
Specifications
Cartridge/FilterPARTICULATE
Certifications/ComplianceNIOSH
ColorWhite
Filter TypeP3 TM3
Product TypeFilter
With Exhalation ValveYes
Facepiece TypeSteri-Plus Case, CS3038
Filtration Layer MaterialNonwoven, Electrostatic Media
For Use With (Equipment)Steri-Plus Reusable Case and CSPAPR
Latex FreeYes
ModelCS3039
Quantity20 Pk.
SterilityNon-sterile
Dimensions (L x W x H)20 x 19 x 3 cm
Unit SizePack