Steri-Plus Exhalation Valve Filter Reusable Case for Source Control
Actual product may vary
CleanSpace
Steri-Plus Exhalation Valve Filter Reusable Case for Source Control
Case is compatible with half masks and utilized for source control in sterile settings.
Have Questions?
Catalog number CSECS3038
Price (SGD)
79.00
Each
Price (SGD)
79.00
Each
The world's first PAPR exhalation valve filter for source control. Developed to filter the air exhaled by the wearer and consequently protect those not wearing a respiratory protection system.
  • Filtration efficiency of 97% for particles 0.3 μm and above
  • Approved for use with CleanSpace™ respirators and half masks
  • Reusable case is compatible with standard disinfection and sterilization protocols
  • Easy to fit and clean
  • Cost-effective
  • NIOSH and CE Mark approved
  • Suitable for use in patient care settings and laboratory or pharmaceutical clean rooms
  • Provides reliable source control for CleanSpace™ respirators when used with the half mask
  • Exhalation valve filter reusable case contains polycarbonate outer case and ABS inner case
Specifications
ColorBlue, Gray
DisposableReusable
MaterialABS, Polycarbonate
Product TypeValve Filter Case
Facepiece TypeAttaches to Exterior of Facemask
For Use With (Application)Source Control
For Use With (Equipment)CleanSpace™ PAPR and Mask
IncludesReusable Case Only
Latex FreeYes
ModelCS3038
SterilityNon-sterile
Length (English)13 in.
Length (Metric)33 cm
Width (English)3.93 in.
Width (Metric)10 cm
Height (English)7.9 in.
Height (Metric)20 cm
Weight (English)2.5 oz.
Weight (Metric)70.5 g
Unit SizeEach