The BioHood System is for use with the HALO and is specifically designed for biocontainment units, laboratories and pharmaceutical cleanrooms.
Have Questions?
Catalog number CSECS3025
Price (SGD)
228.00
Pack
Price (SGD)
228.00
Pack
The BioHood System is compatible with the CleanSpace™ HALO Respirator. It is specifically designed for biocontainment units, laboratories and pharmaceutical cleanrooms, providing eye and head protection.
DuPont™ Tyvek™ material and seam technology
Light and durable
Blood and fluid resistant
Only PAPR hood design to completely cover the respirator when worn, minimizing device contamination
Can be easily removed while still wearing the respirator to protect healthcare professionals when doffing
Broad field of vision and unique design that secures the visor to eliminate slippage and movement
No belts, hoses, fogging or heat
Comfortable air flow with no breathing resistance
NIOSH and CE Mark approved
BioHood Parts: The BioHood (CS3024), Bio HEPA P3/T3 (CS3025), Bio Mask Exhalation Adaptor (CS3026) - reusable and attaches to CleanSpace™ HALO Face Mask (face mask sold separately)
Specifications
Cartridge/FilterPARTICULATE
ColorBlue, Gray
Filter TypeHEPA
Product TypeFilter
With Exhalation ValveNo
Filter EfficiencyP3 TM3 P SL R
For Use With (Equipment)CS3024, CS3025, CS3026, CS3027