BioHood
Actual product may vary
CleanSpace
BioHood
The BioHood System is for use with the HALO and is specifically designed for biocontainment units, laboratories and pharmaceutical cleanrooms.
Have Questions?
Catalog number CSECS3024
Price (SGD)
220.00
Each
Price (SGD)
220.00
Each
The BioHood System is compatible with the CleanSpace™ HALO Respirator. It is specifically designed for biocontainment units, laboratories and pharmaceutical cleanrooms, providing eye and head protection.
  • DuPont™ Tyvek™ material and seam technology
  • Light and durable
  • Blood and fluid resistant
  • Only PAPR hood design to completely cover the respirator when worn, minimizing device contamination
  • Can be easily removed while still wearing the respirator to protect healthcare professionals when doffing
  • Broad field of vision and unique design that secures the visor to eliminate slippage and movement
  • No belts, hoses, fogging or heat
  • Comfortable air flow with no breathing resistance
  • NIOSH and CE Mark approved
  • BioHood Parts: The BioHood (CS3024), Bio HEPA P3/T3 (CS3025), Bio Mask Exhalation Adaptor (CS3026) - reusable and attaches to CleanSpace™ HALO Face Mask (face mask sold separately)
Specifications
ColorWhite
MaterialFlash Spun Polyethylene
Facepiece TypePolycarbonate Visor
For Use With (Application)Protection From Splash and Airborne Contaminents
For Use With (Equipment)CS3025, CS3027
Latex FreeYes
SterilityNon-sterile
Dimensions (L x W x H)31 x 14 x 10 cm
Unit SizeEach