▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Air-Purifying Respirators
▸
Powered Air Purifying Respirators
▸
PAPR Replacement Parts and Accessories
▸
Station Charging and Storage Case
Actual product may vary
CleanSpace
Station Charging and Storage Case
Holds up to 8 units.
Have Questions?
Catalog Number
For Use With (Equipment)
CSECS3014
CS3000 PAPR System
Catalog number
CSECS3014
Price (SGD)
12,873.00
Each
Add to cart
For Use With (Equipment):
CS3000 PAPR System
Price (SGD)
12,873.00
Each
Add to cart
For use with HALO Power Systems
Holds up to 8 units
Specifications
Color
Clear
Disposable
Reusable
Product Type
Station Charger/Case
For Use With (Equipment)
CS3000 PAPR System
Model
CS3014