▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Air-Purifying Respirators
▸
Powered Air Purifying Respirators
▸
PAPR Replacement Parts and Accessories
▸
CleanSpace3™ Head Harness for Half Mask (non fabric)
Actual product may vary
CleanSpace
CleanSpace3™ Head Harness for Half Mask (non fabric)
Replacement part for use with the CleanSpace HALO. Consists of TPU clips, polyethylene strap with polyester adjustment band.
Have Questions?
Catalog Number
For Use With (Equipment)
CSECS3008
CS3000 PAPR System
Catalog number
CSECS3008
Price (SGD)
113.00
Each
Add to cart
For Use With (Equipment):
CS3000 PAPR System
Price (SGD)
113.00
Each
Add to cart
Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
Designed for comfort over long periods
Allows sweating and breathability
Multiple adjustment points
Easy to wash and quick drying
Durable and designed for long wear in harsh environments
Easy and quick replacement
Specifications
Disposable
Reusable
Product Type
Head Harness
For Use With (Equipment)
CS3000 PAPR System
Model
CS3008