HALO Half Mask for PAPR Assembly
Actual product may vary
CleanSpace
HALO Half Mask for PAPR Assembly
Specifically designed to work to complete the CleanSpace HALO system and clip quickly onto the power units. Half masks are made of soft silicone that readily molds to the face and does not cause allergic dermatitis.
Have Questions?
Catalog NumberSize
CSECS3005Large
Catalog number CSECS3005
Price (SGD)
261.00
Each
Size:
Large
Price (SGD)
261.00
Each
  • Used with the revolutionary CleanSpace HALO: light weight, no hoses or belts
  • Half masks available in 3 sizes to facilitate a good seal and a comfortable fit
  • Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
  • Mask Fit Test adaptor available for TSI Portacount™ fit testing
  • Easy removal
  • Compatible with most cleaning and disinfection protocols
  • Transparent and facilitates communication
If you require intrinsically safe equipment, please consult your site Intrinsically Safe approvals. These Half Masks are NOT certified intrinsically safe.
Specifications
Certifications/ComplianceAS/NZS1716: 2012
Certifications/Compliance (Europe)EN 12942
Product TypePAPR Half Mask
SizeLarge
Facepiece/Headtop ModelHalf mask only
For Use With (Application)Primary healthcare facilities, pharmaceutical production, laboratories, animal research facilities and emergency/disaster responders
Intrinsic Safety (IS) RatingNo
ModelCS3005
Unit SizeEach