HALO Particulate Filter P3 Standard
Actual product may vary
CleanSpace
HALO Particulate Filter P3 Standard
Suitable for protection against airborne particulates (dust, mists and fumes) and biohazards.
Have Questions?
Catalog number CSECS3002
Price (SGD)
150.00
Pack
Price (SGD)
150.00
Pack
  • Used with the revolutionary CleanSpace PAPR: light weight, no hoses or belts
  • Suitable for protection against airborne particulate (dust, mists and fumes).
  • Materials: Fiberglass particulate media and plastic casing, silicone seal
  • Easy and quickly fitted and removed from the power unit
  • Pack of 3 (CS3002)
IMPORTANT: The filters are compatible with CleanSpace HALO Power Units. When selecting a CleanSpace Filter please consult a Health and Safety specialist for advice on the appropriate respiratory equipment and filter use.
Specifications
Product TypeFilter
Unit SizePack