HALO Power System
Actual product may vary
CleanSpace
HALO Power System
Battery-powered, positive pressure air-purifying respirator with IP Rating 66. Suitable for healthcare, laboratory, pharmaceutical and emergency response. Protection in applications with particulate contaminants and biohazards as well as heavy water and decontamination procedures.
Have Questions?
Catalog number CSECS3000
Price (SGD)
1,800.00
Special offer
Ends: 31-Dec-2026
1,966.00
Save 166.00 (8%)
Each
Price (SGD)
1,800.00
Special offer
Ends: 31-Dec-2026
1,966.00
Save 166.00 (8%)
Each
  • IP Rating 66 – resistant to dust and liquid ingress
  • Suitable for decontamination procedures
  • Light weight (350g),compact and comfortable
  • Long operating time: up to 9 hours and quick battery recharging: < 2hours
  • AirSensit™ System for mask pressure control and breath responsive airflow
  • One-button system and standby and auto-start/stop mode for quick don/doffing
  • Easy to use support harness
  • Altitude compensated filter blocked detection
  • Easy and accurate flow capability check
Mask sold separately
Specifications
Product TypePAPR Assembly, No Headtop
ModelCS3000
Unit SizeEach