Sentino™ Filters Funnel, Gamma Irradiated
Actual product may vary
Sentino™ Filters Funnel, Gamma Irradiated
Sentino™ Filter Funnels are single-use funnels designed for use with the Sentino™ pump with a press-fit seal, membrane guides and forceps access point allowing for simple set up and membrane removal.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
CRG4871250 mL20/Sleeve, 80/Pk.
CRG4870100 mL25/Sleeve, 100/Pk.
Catalog number CRG4871
Price (SGD)
187.00
Case
Capacity (Metric):
250 mL
Quantity:
20/Sleeve, 80/Pk.
Price (SGD)
187.00
Case
  • For research use only
Specifications
Capacity (Metric)250 mL
MaterialPolypropylene
Quantity20/Sleeve, 80/Pk.
SterilitySterile
Product TypeFiltering Funnel
Unit SizeCase