Check Order Status
Sentino™ Filters Funnel, Gamma Irradiated
Actual product may vary
Sentino™ Filters Funnel, Gamma Irradiated
Sentino™ Filter Funnels are single-use funnels designed for use with the Sentino™ pump with a press-fit seal, membrane guides and forceps access point allowing for simple set up and membrane removal.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Quantity
CRG4870
100 mL
25/Sleeve, 100/Pk.
CRG4871
250 mL
20/Sleeve, 80/Pk.
2 Options
Catalog number
CRG4870
Price (SGD)
124.00
Special offer
Ends:
31-Mar-2026
198.00
Save 74.00 (37%)
CS
Add to cart
Capacity (Metric):
100 mL
Quantity:
25/Sleeve, 100/Pk.
Price (SGD)
124.00
Special offer
Ends:
31-Mar-2026
198.00
Save 74.00 (37%)
CS
Add to cart
For research use only
Specifications
Capacity (Metric)
100 mL
Material
Polypropylene
Quantity
25/Sleeve, 100/Pk.
Sterility
Sterile
Product Type
Filtering Funnel