Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile
Actual product may vary
Corning
Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile
Manufactured with optically clear polystyrene for accurate measurement. Corning™ Stripette™ Individually Wrapped Plastic or Paper/Plastic Disposable Polystyrene Serological Pipets are ideal for sterile tissue culture applications, especially while wearing latex gloves, as the wrap reduces static cling.
Have Questions?
Catalog NumberVolume (Metric)Color
CRG4491100 mLBlue
Catalog number CRG4491
Price (SGD)
638.00
Case
Volume (Metric):
100 mL
Color:
Blue
Price (SGD)
638.00
Case
  • Sterile
  • RNase-/DNase-free
  • Nonpyrogenic
  • Noncytotoxic
  • BSE/TSE-free
  • Exclusive anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Bi-directional graduations provide choice of ascending and descending scales
  • Negative graduations allow additional working volume
  • Three packaging options: Individually wrapped, paper/plastic; Individually wrapped, clear plastic; Bulk packed
  • Volumetric accuracy of ±2% at full capacity
Specifications
Accuracy±2%
Certifications/ComplianceNon-pyrogenic, DNase, RNase-free
MaterialPolystyrene
SterilitySterile
GraduatedYes
Graduations1 mL
Length (English)13.7 in.
Quantity100/Cs.
Tip StyleRound
Tolerance±2%
Packaging TypeIndividually Wrapped, Paper/Plastic Packaging
Volume (English)3.38 oz.
ColorBlue
Volume (Metric)100 mL
Length (Metric)34.8 cm
TypeSerological Pipet