Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Type | Neck Style |
---|---|---|---|
CRG431080 | 750 mL | Vented | Angled |
FIS10-126-28 | 70 mL | Vented | Canted |
FIS10-126-32 | 600 mL | Plug Seal | Canted |
FIS10-126-34 | 600 mL | Vented | Canted |
FIS10-126-62 | 1000 mL | Plug Seal | Angled |
FIS10-126-63 | 1000 mL | Vented | Angled |
Cap Types