Search Fisher Scientific
| Catalog Number | Material (Membrane) | Funnel Capacity | Pore Size |
|---|---|---|---|
| CRG430517 | Cellulose Acetate | 1000 mL | 0.22 μm |
| CRG431474 | PES | 1000 mL | 0.1 μm |
| FIS09-761-180 | PES | 1000 mL | 0.1 μm |
| FKS11363785 | PES | 1000 mL | 0.1 μm |
| FIS09-761-40 | Cellulose Acetate | 1000 mL | 0.22 μm |
| CRG431098 | PES | 1000 mL | 0.22 μm |
| FIS09-761-108 | PES | 1000 mL | 0.22 μm |
| FKS10296191 | PES | 1000 mL | 0.22 μm |
| FIS09-761-104 | Cellulose Nitrate | 1000 mL | 0.22 μm |
| FIS09-761-41 | Cellulose Acetate | 1000 mL | 0.45 μm |
| FKS10730693 | Cellulose Acetate | 1000 mL | 0.45 μm |