Disposable Polystyrene Serological Pipets, Sterile
Actual product may vary
Disposable Polystyrene Serological Pipets, Sterile
Features exceptionally crisp, dark, easy-to-read printing with reverse and negative graduations. Falcon™ Disposable Polystyrene Serological Pipets are individually wrapped.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Graduations
CRG35755050 mL1 mL
CRG3575211 mL0.01 mL
FIS13-675-15B1 mL0.01 mL
FIS13-675-172 mL0.01 mL
CRG3575435 mL0.1 mL
FIS13-675-225 mL0.1 mL
FKS104683125 mL0.1 mL
CRG35755110 mL0.1 mL
FIS13-675-2010 mL0.1 mL
FKS1028237110 mL0.1 mL
CRG35752525 mL0.25 mL
FIS13-668-225 mL0.25 mL
FIS13-675-2750 mL1 mL
FIS13-675-73100 mL1 mL
Catalog number CRG357550
Price (SGD)
112.00
Case
Volume (Metric):
50 mL
Graduations:
1 mL
Price (SGD)
112.00
Case
  • Polystyrene, disposable pipets for tissue culture, bacteriological, and research applications
  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0.1 EU/mL
  • BSE/TSE-free
  • Negative graduations for extra capacity
  • Reverse graduations (except 1 mL)
  • Color-coded package and markings for ease of identification
  • Polyester plug to help prevent overfill
  • Accurate ±2% at full volume (excluding 1 and 2 mL)
Graduation alignment system
The pipet graduation alignment system is a fast and simple way to bring graduations to your line of sight. Simply pop open or peel back the top of the pipet package and line up the pipettor before completely removing the pipet from its wrapping. When you remove the pipet from its package, the graduations should face you every time.
Specifications
Accuracy±3%
Aspirator PlugYes
AutoclavableNot Autoclavable
Certifications/ComplianceNon-pyrogenic, ISO 9001: 2000, FDA, 820 CFR
Coefficient of Variation1.5%
MaterialPolystyrene
SterilitySterile
For Use With (Application)For tissue culture usage or any non toxic solution
GraduatedYes
Graduations1 mL
Graduation TypeNegative, Reverse Graduations
Negative Graduations10 mL
Quantity100/Cs.
Tip StyleStandard
Tolerance±2%
Packaging TypeIndividually Wrapped, Paper-Plastic
Volume (English)1.6 oz.
ColorBlack
Volume (Metric)50 mL
TypeSerological Pipet