Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Graduations |
---|---|---|
CRG357543 | 5 mL | 0.1 mL |
CRG357521 | 1 mL | 0.01 mL |
FIS13-675-15B | 1 mL | 0.01 mL |
FIS13-675-17 | 2 mL | 0.01 mL |
FIS13-675-22 | 5 mL | 0.1 mL |
FKS10468312 | 5 mL | 0.1 mL |
CRG357551 | 10 mL | 0.1 mL |
FIS13-675-20 | 10 mL | 0.1 mL |
FKS10282371 | 10 mL | 0.1 mL |
CRG357525 | 25 mL | 0.25 mL |
FIS13-668-2 | 25 mL | 0.25 mL |
CRG357550 | 50 mL | 1 mL |
FIS13-675-27 | 50 mL | 1 mL |
FIS13-675-73 | 100 mL | 1 mL |
Graduation alignment system
The pipet graduation alignment system is a fast and simple way to bring graduations to your line of sight. Simply pop open or peel back the top of the pipet package and line up the pipettor before completely removing the pipet from its wrapping. When you remove the pipet from its package, the graduations should face you every time.