BioCoat™ Collagen I Multiwell Plates
BioCoat™ Collagen I Multiwell Plates
Actual product may vary
BioCoat™ Collagen I Multiwell Plates
Treated with BioCoat™ for enhanced cell attachment and growth of a variety of primary cells and ransformed cell lines in serum-free or reduced serum cultures. Corning™ BioCoat™ Collagen I Multiwell Plates closely mimmics in vivo environments for cell culture applications. Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
Have Questions?
Catalog NumberCulture Area
CRG3564009.6 cm2
Catalog number CRG356400
Price (SGD)
1,157.00
CS
Add to cart
Culture Area:
9.6 cm2
Price (SGD)
1,157.00
CS
Add to cart
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
Applications include:
  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture
Source
  • Rat tail tendon
  • Quality Control
    • Tested for ability to promote attachment and spreading of HT-1080 human fibrosarcoma cells
    • Tested and found negative for bacteria and fungi
    • Collagen I purity >90% by SDS-PAGE
    Storage
  • 4°C to 30°C under dry conditions.
  • Specifications
    Array2 x 3
    Certifications/ComplianceANSI, SBS
    Closure ColorClear
    Closure IncludedYes
    Closure MaterialPolystyrene
    Bottom ShapeFlat
    ColorClear
    Culture Area9.6 cm2
    MaterialPolystyrene
    ShapeRectangular
    SterilityNon-sterile
    Surface TreatmentCell Culture
    TypeMulti-well Cell Culture Plate
    Well Count6
    For Use With (Application)Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
    IncludesLid
    No. per Case50/Cs.
    No. per Pack5/Pk.
    Packaging10 inner bags of 5 each
    Quantity50/Cs.
    Shelf Life6 Months
    SourceRat Tail
    Storage Requirements4°C to 30°C
    Unit SizeCS