Specifications
Array4 x 6
Closure ColorClear
Closure IncludedYes
Closure MaterialPolystyrene
Bottom ShapeFlat
Capacity (Metric)3.5 mL
ColorClear
Culture Area2 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
Surface TreatmentCell Culture
TypeCell Culture Plate
Volume (Metric) Working1.5 to 2.2 mL
Well Count24
IncludesLid
No. per Case50/Cs.
No. per Pack1/Pk.
PackagingIndividually Wrapped
Pore Size1 μm
Quantity50/Cs.
Diameter (Metric)6.5 mm
Unit SizeCase