Round-Bottom Polystyrene Test Tubes with Cell Strainer Snap Cap, 5mL
Designed for flow cytometry applications. Falcon™ 12x75mm Test Tube with Cell Strainer Snap Cap offer a convenient solution to prepare laboratory samples.
Have Questions?
Catalog Number
Includes
CRG352235
35 μm Nylon Mesh Screen
Catalog number CRG352235
Price (SGD)
1,159.00
Case
Includes:
35 μm Nylon Mesh Screen
Price (SGD)
1,159.00
Case
Widely used and referenced in laboratory protocols
Dual position snap-cap prevents sample loss with ease of handling
Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352008 nonsterile)
The foundation for consistent research results
Provide reliable containment of laboratory fluid samples
Polystyrene tubes are best suited for procedures requiring high optical clarity
Dual-position snap caps, heavier gauge walls, and unique construction provide a secure, positive seal
Medical-style packaging materials
1,400 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
Tube dimensions and volumes are approximate
OD: 12 mm
Graduations: No
Specifications
ColorClear
Capacity (Metric)5 mL
Length (Metric)75 mm
MaterialPolystyrene
For Use WithCorning FACS System
Includes35 μm Nylon Mesh Screen
SterilitySterile, nonpyrogenicity tested to less than 0.1 EU/ mL