50 mL High Clarity Conical Centrifuge Tubes
Actual product may vary
50 mL High Clarity Conical Centrifuge Tubes
Tested to withstand centrifugation of 16,000 RCF in a fully supported rotor with room temperature water. Falcon 50mL Conical Centrifuge Tubes are ideal for cell centrifugation; pelleting; separation by density gradients.
Have Questions?
Catalog NumberPackagingMax. RCF
CRG352098Rack16,000 x g
Catalog number CRG352098
Price (SGD)
162.00
Case
Packaging:
Rack
Max. RCF:
16,000 x g
Price (SGD)
162.00
Case
Suitable for molecular biology applications including concentrating bacteria for DNA isolation, purification and precipitation of nucleic acids; sample storage (ambient temperature to -80°C); and centrifugation of precipitates. 
  • Hydrophobic, biologically inert surface for good cell or protein recovery
  • Dark blue graduations in 5mL increments
  • White writing area resists solvents
  • Irradiated for sterility
 Clarified Polypropylene
  • Resistant to alcohols and mild organic solvents (not recommended for extraction procedures)
  • Translucent wall for easy viewing
  • Stable from -80°C to +121°C (-112°F to +250°F)
  • Withstand centrifugal forces to 16,000 x g RCF
Specifications
Certifications/ComplianceISO 9001:2000, FDA, CFR
Closure ColorBlue
Diameter (English) Outside1.2 in.
ColorClear
Capacity (Metric)50 mL
MaterialPolypropylene
For Use WithFor DNA isolation, purification and precipitation of nucleic Acids, cell centrifugation applications
No. per Case500
PackagingRack
SterilitySterile
ShapeConical
Length (Metric)115 mm
Capacity (English)1.69 oz.
Bottom ShapeConical
Closure TypeFlat Top with Rim Seal
Writing AreaWhite Marking Spot
Diameter (Metric)30 mm
GraduatedYes
Product TypeCentrifuge Tube
Max. RCF16,000 x g
Unit SizeCase