Manufactured from polypropylene for excellent chemical and thermal resistance. Falcon™Round-Bottom Polypropylene Tubes are tested to withstand 3000 x g centrifugation.
Have Questions?
Catalog Number
Capacity (Metric)
Length (Metric)
Packaging
CRG352059
14 mL
100 mm
25/Pk.; 500/Cs.
Catalog number CRG352059
Price (SGD)
118.00
Special offer
Ends: 31-Dec-2026
194.00
Save 76.00 (39%)
Case
Capacity (Metric):
14 mL
Length (Metric):
100 mm
Packaging:
25/Pk.; 500/Cs.
Price (SGD)
118.00
Special offer
Ends: 31-Dec-2026
194.00
Save 76.00 (39%)
Case
Widely used and referenced in laboratory protocols
Dual position snap-cap prevents sample loss with ease of handling
Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352002 nonsterile)
The foundation for consistent research results
Provide reliable containment of laboratory fluid samples
Polypropylene tubes are best suited for applications requiring greater thermal and chemical stability
Dual-position snap caps, heavier gauge walls, and unique construction provide a secure, positive seal
Medical-style packaging materials
3,000 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
17 x 100 mm tubes have blue printed graduations and white writing patch