Round-Bottom Polystyrene Test Tubes
Round-Bottom Polystyrene Test Tubes
Actual product may vary
Round-Bottom Polystyrene Test Tubes
Constructed of optically clear polystyrene for easy viewing of sample. Falcon™ Round-Bottom Polystyrene Tubes are widely used and referenced in laboratory protocols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure TypeSterilityPackaging
CRG3520545 mLSnap CapSterileBag of 125, 1000/Cs.
CRG35204519 mLScrew CapSterileIndividually wrapped, 500/Cs.
FIS14-959-40B19 mLScrew CapSterileIndividually wrapped, 500/Cs.
FKS1069433119 mLScrew CapSterileIndividually wrapped, 500/Cs.
FIS14-959-4216 mLScrew CapSterileIndividually wrapped, 500/Cs.
FIS14-959-38B16 mLScrew CapSterileBag of 125, 1000/Cs.
FIS14-959-55 mLNot IncludedNon-sterileBulk, 1000/Cs.
FIS14-959-1A5 mLSnap CapSterileBag of 25, 500/Cs.
FIS14-959-1B14 mLSnap CapSterileBag of 25, 500/Cs.
FIS14-959-2A5 mLSnap CapSterileBag of 125, 1000/Cs.
FIS14-959-38C8 mLScrew CapSterileBag of 125, 1000/Cs.
CRG3520525 mLNot IncludedSterile1000/Cs.
FIS14-959-65 mLNot IncludedSterile1000/Cs.
FKS105795115 mLNot IncludedSterile1000/Cs.
FIS14-959A5 mLSnap CapSterileIndividually wrapped, 500/Cs.
FIS14-959B14 mLSnap CapSterileIndividually wrapped, 500/Cs.
Catalog number CRG352054
Price (SGD)
183.00
Special offer
Ends: 31-Mar-2026
273.00
Save 90.00 (33%)
CS
Add to cart
Capacity (Metric):
5 mL
Closure Type:
Snap Cap
Sterility:
Sterile
Packaging:
Bag of 125, 1000/Cs.
Price (SGD)
183.00
Special offer
Ends: 31-Mar-2026
273.00
Save 90.00 (33%)
CS
Add to cart
  • Widely used and referenced in laboratory protocols
  • Dual position snap-cap prevents sample loss with ease of handling; provides a secure, positive seal (except for items 352008, 352017, and 352052 which do not have caps) 
  • Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352008 nonsterile)
The foundation for consistent research results
  • Provide reliable containment of laboratory fluid samples
  • Polystyrene tubes are best suited for procedures requiring high optical clarity
  • Heavier gauge walls; unique construction 
  • Medical-style packaging materials
  • 1,400 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
  • Tube dimensions and volumes are approximate
Specifications
Closure ColorNatural
Closure MaterialPolyethylene
ColorClear
Description1400 x G
Capacity (Metric)5 mL
Length (Metric)75 mm
MaterialPolystyrene
DisposableSingle-use
IncludesDual position snap cap
PackagingBag of 125, 1000/Cs.
SterilitySterile
GraduatedNo
Diameter (Metric)12 mm
Closure TypeSnap Cap
Product TypeTest Tube
Unit SizeCS