Costar™ 6-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
Costar™ 6-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
Actual product may vary
Costar™ 6-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
Have Questions?
Catalog NumberDiameter (Metric)
CRG35163.5 cm
Catalog number CRG3516
Price (SGD)
80.00
Special offer
Ends: 31-Mar-2026
133.00
Save 53.00 (40%)
CS
Add to cart
Diameter (Metric):
3.5 cm
Price (SGD)
80.00
Special offer
Ends: 31-Mar-2026
133.00
Save 53.00 (40%)
CS
Add to cart
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Specifications
Array2 x 3
Closure ColorClear
Closure IncludedYes
Closure MaterialPolystyrene
Bottom ShapeFlat
Capacity (Metric)2.9 mL (Well)
ColorClear
Culture Area9.5 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
Surface TreatmentCell Culture
TypeMulti-well Cell Culture Plate
Volume (Metric) Working2 to 3 mL
Well Count6
IncludesLid
No. per Case50/Cs.
No. per Pack1/Pk.
PackagingIndividually Wrapped
Quantity50/Cs.
Diameter (Metric)3.5 cm
Unit SizeCS