Search Thermo Fisher Scientific
Catalog Number | Well Shape | Surface Treatment | Sterility | Lid |
---|---|---|---|---|
CRG3370 | Flat Bottom (F-well) | Non-treated | Sterile | With Lid |
FIS07-200-107 | Conical Bottom (V-well) | Non-treated | Sterile | Without Lid |
FKS10296381 | Round | Non-treated | Sterile | Without Lid |
FIS07-200-108 | Conical Bottom (V-well) | Non-treated | Non-sterile | Without Lid |
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.