Sterile Spatulas, small spoon
Actual product may vary
Corning
Sterile Spatulas, small spoon
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
Have Questions?
Catalog NumberLength (Metric)
CRG300423.4 cm
Catalog number CRG3004
Price (SGD)
318.00
Special offer
Ends: 06-Sep-2026
523.00
Save 205.00 (39%)
Case
Length (Metric):
23.4 cm
Price (SGD)
318.00
Special offer
Ends: 06-Sep-2026
523.00
Save 205.00 (39%)
Case
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
  • Corning spatulas are designed to save time and to provide contamination-free samples.
  • Eliminates the recycling and resterilizing necessary with reusable spatulas.
  • Available in five different configurations.
  • Microspatulas available in two configurations.
  • Individually packaged.
  • Antistatic.
  • Sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Specifications
SterilitySterile
ShapeSpoon
Certifications/ComplianceUSP Class VI, non-pyrogenic, RNase and DNase-free Certified
DisposableSingle-use
Product TypeSpatula, Spatula
For Use With (Application)Molecular biology applications, lyophilized samples and contamination-free sampling
Length (English)9.25 in.
IncludesNarrow small spoon at one end and a larger round spoon at the other end
MaterialPolystyrene
Length (Metric)23.4 cm
Unit SizeCase