HydroPure™ with Purified Water
Actual product may vary
Contec
HydroPure™ with Purified Water
HydroPure™ with Purified Water designed for cleaning of life science cleanrooms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberContainer TypeQuantity
CNTSBT16HPWTrigger Spray Bottle6 x 1 L
CNTSBC56HPCapped Bottle2 x 5 L
Catalog number CNTSBT16HPW
Price (SGD)
426.00
Case
Container Type:
Trigger Spray Bottle
Quantity:
6 x 1 L
Price (SGD)
426.00
Case
Designed for cleaning of life science cleanrooms.
  • Composition: 6% hydrogen peroxide in water
  • Can be applied directly to surface ensuring even coverage
  • Filtered to 0.22 microns under Grade A airflow
  • Physically removes contaminants
  • Sufficient sporicidal activity for Grade A / B area
  • Low residue, eliminates need for residue removal stage with alcohol or water
  • Gamma irradiated
  • Large droplet size
  • Facilitates transfer disinfection into isolators and RABS
  • Same grade as when vaporized hydrogen peroxide is used
  • Triple / double bagged in linear tear packaging
  • Noncorrosive
Applications:
  • Efficacious against bacteria, fungi, moulds, yeasts and spores
Specifications
Product TypeDetergent with Purified Water
FormLiquid Spray
pH6.5 to 7.5
SterilitySterile
AppearanceColorless
AutoclavableNot Autoclavable
ColorClear
CompositionHydrogen peroxide and purified water (EP)
Concentration6% Hydrogen peroxide in purified water (EP)
Container TypeTrigger Spray Bottle
DisposableLimited-use
FilteredFiltered
FlammableNo
For Use With (Application)Used in pharmaceutical industry
Quantity6 x 1 L
Shelf Life1 Year
Volume (English)33.81 oz.
Volume (Metric)1 L
Unit SizeCase