ProChlor
Actual product may vary
Contec
ProChlor
Contec ProChlor is a revolutionary new sporicide achieving a 100% kill against spores in under 1 min.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberContainer TypeQuantitySterility
CNTSBC502PCCapped Bottle2 x 5 LSterile
CNTSBT102PCTrigger Spray Bottle6 x 1 LSterile
Catalog number CNTSBC502PC
Price (SGD)
476.00
Case
Container Type:
Capped Bottle
Quantity:
2 x 5 L
Sterility:
Sterile
Price (SGD)
476.00
Case
Ensures cleanroom is free from contamination and particulates.
A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts, viruses and spores.
Contec ProChlor is 0.2 micron filtered, and filled in a Grade A environment. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.
Features & Benefits
  • Log 6 kill against Bacillus subtilis in 60 secs saves time spent on biodecontamination
  • Filtered to 0.2 microns in a Grade B cleanroom ensures the product is free from contamination and particulates
  • Sterile version available for Grade A and B cleanrooms
  • Contains no quaternary ammonium or surfactant so it's very low residue
  • No hazard classification means only basic PPE required and no special disposal is required
  • Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
  • Not classed as corrosive and can be used safely in all areas of the cleanroom
Note: For trigger spray option, see 15567161.
Specifications
Product TypeDisinfectant
FormLiquid
pH6.5 to 7.5
SterilitySterile
AppearanceColorless
AutoclavableNot Autoclavable
ColorClear
CompositionStabilized Hypochlorous acid blended with purified water
Concentration2000 ppm hypochlorous acid
Container TypeCapped Bottle
DisposableLimited-use
FilteredFiltered
FlammableNo
For Use With (Application)Ready-to-Use Hard Surface Disinfectant
PackagingDouble packed in polyethylene bags
Quantity2 x 5 L
Shelf Life2 Years
Volume (English)169.07 oz.
Volume (Metric)5 L
Unit SizeCase
Contents & Storage
Do not freeze. Store below 40°C.