Sterile 70% Isopropanol
Sterile 70% Isopropanol
Actual product may vary
Sterile 70% Isopropanol
Contec™ Sterile 70% Isopropanol is ideal for wiping down of isolators, cabinets, laminar flow hoods and surfaces.
Have Questions?
Catalog NumberVolume (English)
CNTSB167030IR16 oz.
Catalog number CNTSB167030IR
Price (SGD)
-
Volume (English):
16 oz.
Contains 70% by volume USP-grade Isopropanol (IPA) and 30% USP-purified water.
  • Evaporates and leaves extremely low residues
  • Eliminates mixing, filtration, processing and QC expense of in-house production of sterile isopropanol water blends
  • Filtered through 0.2μm in-line filter which eliminates the need for in-house filtration
  • Gamma-irradiated to a sterility assurance level (SAL) of 10-6
  • Prefiltered, packaged and sterilized for easy introduction into critical environments
  • Certificate of processing and sterility with each case
  • Induction-sealed bottles reduce opportunity for leakage during sterilization and shipment
  • 16 and 32 oz. sizes include a flexible trigger sprayer for instant adjustment for a variety of applications
  • Recyclable plastic bottle
Specifications
FormLiquid
SterilitySterile
AppearanceClear
Certifications/ComplianceUSP 797
Composition70% USP Isopropyl Alcohol and 30% USP Purified Water
For Use With (Application)Wipe down of isolators, cabinets, laminar flow hoods and surfaces
IncludesPlastic bottle with trigger sprayer
Volume (English)16 oz.
Volume (Metric)0.47 L
Unit SizeCS