Amplitude™ Kappa™ Sterile LE Wipes
Amplitude™ Kappa™ Sterile LE Wipes
Actual product may vary
Amplitude™ Kappa™ Sterile LE Wipes
Designed for wiping in ISO Class 5 environments. Contec™ Amplitude™ Kappa™ Sterile LE Wipes are low endotoxin wipes that meet the FDA standard for bacterial endotoxins using Limulus Amoebocyte Lysate at the endotoxin limit for medical devices at <20 EU/device.
Have Questions?
Catalog NumberDimensions (L x W)
CNTNWPZ00019 x 9 in. (22.86 x 22.86 cm)
Catalog number CNTNWPZ0001
Price (SGD)
640.00
Special offer
918.00
Save 278.00 (30%)
EA
Add to cart
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Price (SGD)
640.00
Special offer
918.00
Save 278.00 (30%)
EA
Add to cart
  • Sterile, nonpyrogenic nonwoven wiper made of cellulose-free lyocell (rayon) and polyester material
  • Ideal for product wipe down and cleaning the exterior surfaces of equipment in the biotech, pharmaceutical, and medical device industries
  • Unique surface texture provides effective cleaning and particle removal
  • Low levels of particles and extractables
  • High water and solvent sorbent capacity and rate
  • Gamma-irradiated to a Sterility Assurance Level (SAL) of 10-6
Specifications
Product TypeCleanroom Wipe
Length (English)9 in.
Length (Metric)23 cm
Width (English)9 in.
Width (Metric)23 cm
MaterialLyocell, Polyester
Certifications/ComplianceISO Class 5
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)Ideal for wiping down products and cleaning equipment surfaces in sterile areas