Quiltec™ I Mop Head
Quiltec™ I Mop Head
Actual product may vary
Quiltec™ I Mop Head
Contec Quiltec I Mop Heads are manufactured from 100% 2-ply polyester around a rayon core so are ideal for even the highest grade of cleanroom as they generate very small numbers of particle and fibres.
Have Questions?
Catalog NumberSterility
CNTMHQT0150Sterile
Catalog number CNTMHQT0150
Price (SGD)
351.00
CS
Add to cart
Sterility:
Sterile
Price (SGD)
351.00
CS
Add to cart
  • Two-ply 100% knitted polyester over a rayon core
  • Gamma irradiated at no less than 25 kGy
  • 5 mops double bagged
  • Approx 18-20 sqm coverage
  • Certificates of conformity supplied with every batch
Specifications
Color CodeWhite
Dimensions39 x 13 cm
Product TypeMop Head
SterilitySterile
AutoclavableNot Autoclavable
DisposableSingle-use
For Use With (Application)Cleanroom Mopping
Color-codedNo
MaterialPolyester, Rayon
Unit SizeCS