Check Order Status
GC Stationary Phase, OV-17
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
GC Stationary Phase, OV-17
Suitable for the separation of drugs, amino acid derivatives, pesticides, phenols, and sugar derivatives
Have Questions?
Contact Us
Catalog number
CNT21105
Price (SGD)
236.00
CS
Add to cart
Price (SGD)
236.00
CS
Add to cart
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase
Specifications
Max. Temperature
375°C
Stationary Phase
OV-17
Packaging
Bottle
Quantity
25 g
Description
Chloroform: Soluble
For Use With (Application)
Suitable for G3
Unit Size
CS
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link