Essentials Disposable Plastic Lab Forceps
Actual product may vary
Essentials Disposable Plastic Lab Forceps
Essentials Disposable Plastic Lab Forceps
Essentials Disposable Plastic Lab Forceps
Cole-Parmer
Essentials Disposable Plastic Lab Forceps
The corrugated handles of these plastic forceps provide a secure finger grip for better control.
Have Questions?
Catalog NumberLength (Metric)
CLP06443-2012.2 cm
Catalog number CLP06443-20
Price (SGD)
509.00
Pack
Length (Metric):
12.2 cm
Price (SGD)
509.00
Pack
  • Being disposable reduces cross contamination
  • Serrated tips for accurate sample grip
  • Sterile and individually bagged
  • All plastic forceps have corrugated handles to provide you with a secure finger grip
  • Use serrated pointed tips for handling smaller objects or serrated blunt tips for general use
Specifications
Product TypeForceps
Length (English)4.8 in.
Length (Metric)12.2 cm
MaterialPolystyrene
Quantity100 Forceps/Cs.
SterilitySterile
StyleMicro Alligator Ear
Tip StyleSerrated
Unit SizePack