Clear C-Flex Tubing
Clear C-Flex Tubing
Actual product may vary
Clear C-Flex Tubing
Clear C-Flex Tubing
Cole-Parmer
Clear C-Flex Tubing
Use this tubing for pharmaceutical, biotech and general-purpose lab uses requiring high performance. Saint Gobain Performance Plastics™ Clear C-Flex Tubing features excellent chemical resistance, low gas permeability and a smooth surface.
Have Questions?
Catalog NumberDiameter (Metric) OuterDiameter (Metric) Inner
CLP06422-1111.2 mm6.4 mm
Catalog number CLP06422-11
Price (SGD)
-
Diameter (Metric) Outer:
11.2 mm
Diameter (Metric) Inner:
6.4 mm
Use this tubing for pharmaceutical, biotech and general-purpose lab uses requiring high performance. Saint Gobain Performance Plastics™ Clear C-Flex Tubing features excellent chemical resistance, low gas permeability and a smooth surface.
  • Transparent tubing
  • Economical alternative to platinum silicone
  • Exceptional tensile and tear strength, good biocompatibility
  • Product is manufactured in accordance with Part 820 Quality System Requirements (GMP)
  • Meets USP Class VI
  • Sterilize by autoclave, ethylene oxide or gamma irradiation
Specifications
Certifications/ComplianceUSP, GMP, CFR Title 21, Part 820 Quality System Requirements (GMP)
AutoclavableAutoclavable
Diameter (Metric) Inner6.4 mm
MaterialC-Flex
Diameter (Metric) Outer11.2 mm
Overall Length (Metric)7.62 m
Thickness (Metric)2.38 mm
For Use With (Application)Pharmaceutical, biotech, and general-purpose lab uses requiring high performance, Economical alternative to platinum silicone
Min. Operating Temperature (Metric)-45°C
Packaging Quantity1 Pack
SterilitySterile
ColorTranslucent
Maximum Pressure28 psi
Max. Operating Temperature (Metric)135°C
Unit SizeEA