Transfer Tubing, C-Flex™, Clear
Transfer Tubing, C-Flex™, Clear
Actual product may vary
Transfer Tubing, C-Flex™, Clear
Meets the demands of various industries
Have Questions?
Catalog NumberDiameter (Metric) InnerDiameter (Metric) Outer
CLP06422-053.2 mm6.4 mm
Catalog number CLP06422-05
Price (SGD)
-
Diameter (Metric) Inner:
3.2 mm
Diameter (Metric) Outer:
6.4 mm
Meets the demands of various industries
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Specifications
Certifications/ComplianceUSP Class VI requirements, FDA, GMP
AutoclavableAutoclavable
Diameter (Metric) Inner3.2 mm
MaterialSilicone
Diameter (Metric) Outer6.4 mm
Overall Length (Metric)7.5 m
Thickness (Metric)1.6 mm
For Use With (Application)Pharmaceutical, biotech, and general-purpose lab uses requiring high performance, economical alternative to platinum Silicone
Min. Operating Temperature (Metric)-45°C
Packaging Quantity1 Each
SterilitySterile
ColorTransparent
FlexibilityFlexible
Maximum Pressure35 psi
Max. Operating Temperature (Metric)135°C
Unit SizeEach