1x1 image pixel for data collection
Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Actual product may vary
Arctic Express™ Dual Storage Systems
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
Have Questions?
Catalog NumberCapacity (Metric)
CK5092010 L
Catalog number CK50920
Price (SGD)
4,860.00
EA
Add to cart
Capacity (Metric):
10 L
Price (SGD)
4,860.00
EA
Add to cart
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
  • Warranty varies by country, please contact us for details
Specifications
TypeManual Fill LN2 Dewar
Capacity (Metric)10 L
No. of Doors1
ModelArctic Express Dual 10
Static Holding Time21 shipper/42 lab
Diameter (English)10.2 in.
Diameter (English) Inside Neck2 in.
Diameter (Metric)25.9 cm
Diameter (Metric) Inside Neck5.08 cm
Vial Capacity150 x 2mL
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link