CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up, 797 L
CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up, 797 L
Actual product may vary
CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up, 797 L
CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up, 797 L
CryoExtra™ High-Efficiency Cryogenic Storage Systems with Battery Back-up, 797 L
The Thermo Scientific™ CryoExtra™ Series features automated temperature monitoring, microprocessor-based LN2 level control, and a built-in battery back-up systems, providing peace-of-mind for valuable samples.
Have Questions?
Catalog NumberCapacity (Metric)
CE8140BB797 L
Catalog number CE8140BB
Price (SGD)
92,638.00
EA
Add to cart
Capacity (Metric):
797 L
Price (SGD)
92,638.00
EA
Add to cart
Designed for both liquid and vapor phase storage, the Thermo Scientific™ CryoExtra™ Series features automated temperature monitoring, microprocessor-based LN2 level control, and a built-in battery back-up systems, providing peace-of-mind for valuable samples.
Specifications
TypeAuto-fill LN2 Cryogenic Storage
Capacity (English)28.14 cu. ft.
Capacity (Metric)797 L
No. of Doors1
Cabinet MaterialStainless Steel
InsulationVacuum
Exterior FinishStainless Steel
Certifications/ComplianceCE
Casters3 in. casters (standard)
Dimensions (L x W x H) Exterior49 x 51 x 67 in. (1245 x 1295 x 1702 mm)
Diameter (English)42 in.
Diameter (English) Inside Neck17.5 in.
Diameter (Metric)1067 mm
Diameter (Metric) Inside Neck445 mm
Monitoring OptionsAccess Port for Independent Probe
Amperage0.73A at 110 V and .35A at 230 V
Voltage100/230 V
Electrical Requirements100-230 V
Frequency50/60 Hz
Vial Capacity40,600 2 mL vial capacity (max)
Shipping Weight (Metric)469 kg
Weight (English)720 lb.
Weight (Metric)327 kg
HoldsVials (40,600 x 2 mL)
Unit SizeEA