▸
Solvents
▸
Ethanol
▸
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
Element Search
Structure Search
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
Meets ACS Specifications
Have Questions?
Change view
Quantity:
500 mL
100 mL
2.5 L
4 L
Catalog number
BFRBP2818-500
Price (SGD)
138.00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
138.00
Each
Add to cart
Specifications
Chemical Name or Material
Ethanol
Melting Point
-114°C
Name Note
Absolute (200 Proof)
CAS
64-17-5
ChemAlert Storage Symbol
Red
View more
Meets ACS Specifications
Meets ACS Specifications
For nucleic acid purification and precipitation
Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding