Lindberg/Blue M™ Multipurpose 1500°C Box Furnaces
Lindberg/Blue M™ Multipurpose 1500°C Box Furnaces
Actual product may vary
Lindberg/Blue M™ Multipurpose 1500°C Box Furnaces
Lindberg/Blue M multipurpose furnaces feature integral control to 1500°C. Double-wall insulated construction provides rapid heatup/cooldown and energy efficiency.
Have Questions?
Catalog NumberDescriptionCapacity (Metric)Temperature Control
BF51433PBC-1Box Furnace6 L300 Seg/30 Prog/OTC
Catalog number BF51433PBC-1
Price (SGD)
46,016.00
EA
Add to cart
Description:
Box Furnace
Capacity (Metric):
6 L
Temperature Control:
300 Seg/30 Prog/OTC
Price (SGD)
46,016.00
EA
Add to cart
Thermo Scientific™ Lindberg/Blue M Multipurpose 1500°C Box Furnaces have double-wall construction with Moldatherm™ insulation for rapid heatup and cooldown, energy efficiency and cooler exterior surface temperatures.
Not available in Europe
  • Built-in adjustable high limit overtemperature protection
  • Simultaneous LED display of actual and setpoint temperatures in either °C or °F
  • Silicon carbide heating elements for long-life, safety and reliable service with maximum energy savings
  • Safety door switch interrupts power to heating elements when door is opened; protects elements and minimizes exposure to operator
  • Moldatherm hearthplate supports load and protects interior from spillage and mishandling
  • Type “R” thermocouple is integrated into chamber backwall
With Integral Controller
  • Microprocessor-based PID control
  • Single program with multiple segments for ramp (up and down) and dwell (timed hold) temperature control
With Independent Controller (CC58125C, available separately)
  • Microprocessor-based PID (proportional, integral, derivative) control
  • Single segment, single setpoint, one ramp to setpoint
Requires:
BF51333C requires CC58125C controller
Specifications
Capacity (English)0.21 cu.ft.
DescriptionBox Furnace
Capacity (Metric)6 L
Temperature (Metric)1500°C
TypeMultisegment, Multiprogram
Power Consumption6400 w
Temperature Control300 Seg/30 Prog/OTC
Dimensions (D x W x H) Interior12 x 6 x 5 in. (30.5 x 15.2 x 12.7 cm)
Dimensions (L x W x H) Exterior29 x 25 x 26 in. (73.7 x 63.5 x 66 cm)
Length (English)29 in.
Length (Metric)73.7 cm
Width (English)6 in.
Width (Metric)63.5
Width (Metric) Interior15.2 cm
Height (English)26 in.
Height (English) Interior5 in.
Height (Metric)66 cm
Height (Metric) Interior12.7 cm
Depth (English) Interior12 in.
Depth (Metric) Interior30.5 cm
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V