Sterile SatPax™ 550
Sterile SatPax™ 550
Actual product may vary
Sterile SatPax™ 550
Sterile SatPax™ 550
Sterile SatPax™ 550
Clean 100% melt blown polypropylene pre-wetted wipes provide cost effective and easy to use solution versus traditional bulk handling of solvents, squirt bottles and dry wipes
Have Questions?
Catalog NumberDimensions (L x W)
BERSSP5500042411 x 9 in.
Catalog number BERSSP55000424
Price (SGD)
-
Dimensions (L x W):
11 x 9 in.
  • 100% melt blown polypropylene
  • Solution: 70% IPA / 30% DI water
  • Excellent wet strength and abrasion resistance
  • Gamma irradiated and sterile validated wipes
  • Re-sealable solvent resistant packaging
Specifications
Product TypePre-saturated Cleanroom Wipe
Length (English)11 in.
Length (Metric)28 cm
Width (English)9 in.
Width (Metric)23 cm
MaterialMeltblown Polypropylene
Certifications/ComplianceGamma irradiated and sterile validated to a 10-6 Sterility Assurance Level per AAMI guidelines
ColorWhite
DisposableSingle-use
Dimensions (L x W)11 x 9 in.
For Use With (Application)Final cleaning of surfaces or products prior to manufacturing or packaging
Packaging Quantity50 Sheets/Pk., 24 Pk./Cs.
Saturation Solution70% IPA/30% DI water
SterilitySterile
Unit SizeCS
Contents & Storage
Store at ambient temperature